ENA346974727 Enamine

3,4-bis(trifluoromethyl)pyrrolidine hydrochloride

  • MDL number MFCD20731196
  • Cas Number : 1354959-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95926
Availability details for
Quantity
  • Availability
  • Add to Cart