ENA346974728 Enamine

4-(2-aminoethyl)-1lambda6-thiane-1,1-dione hydrochloride

  • MDL number MFCD20731198
  • Cas Number : 1354951-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95963
Availability details for
Quantity
  • Availability
  • Add to Cart