ENA346974729 Enamine

3-[(2,5-difluorophenyl)methyl]azetidine hydrochloride

  • MDL number MFCD20731199
  • Cas Number : 1354953-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95974
Availability details for
Quantity
  • Availability
  • Add to Cart