ENA346974737 Enamine

2-amino-1-(2-chloro-6-fluorophenyl)propan-1-ol hydrochloride

  • MDL number MFCD20731209
  • Cas Number : 1354949-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96201

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96201
Availability details for
Quantity
  • Availability
  • Add to Cart