ENA346974740 Enamine

2-(4-aminopiperidin-1-yl)pyridine-4-carboxylic acid dihydrochloride

  • MDL number MFCD20731212
  • Cas Number : 1354952-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-96205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96205
Availability details for
Quantity
  • Availability
  • Add to Cart