ENA346974744 Enamine

[(5-cyclopropyl-4-methyl-4H-1,2,4-triazol-3-yl)methyl](methyl)amine

  • MDL number MFCD20731216
  • Cas Number : 1354952-09-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96267
Availability details for
Quantity
  • Availability
  • Add to Cart