ENA346974746 Enamine

2-(4-fluorophenyl)-2-methoxyacetonitrile

  • MDL number MFCD20731218
  • Cas Number : 1354961-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96270

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96270
Availability details for
Quantity
  • Availability
  • Add to Cart