ENA346974750 Enamine

5-(chloromethyl)-3-[1-(2-methylpropoxy)ethyl]-1,2,4-oxadiazole

  • MDL number MFCD20731223
  • Cas Number : 1354949-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96402

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96402
Availability details for
Quantity
  • Availability
  • Add to Cart