ENA346974753 Enamine

3-(1-methyl-1H-1,3-benzodiazol-2-yl)aniline hydrochloride

  • MDL number MFCD20731226
  • Cas Number : 1354952-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96429

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96429
Availability details for
Quantity
  • Availability
  • Add to Cart