ENA346974759 Enamine

3-(5-methyl-1H-1,2,4-triazol-3-yl)propan-1-amine hydrochloride

  • MDL number MFCD20731234
  • Cas Number : 1354953-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96537

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-96537
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart