ENA346974760 Enamine

5-(3-aminopropyl)-2,3-dihydro-1H-1,2,4-triazol-3-one hydrochloride

  • MDL number MFCD20731235
  • Cas Number : 1354953-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231...233
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96538

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96538
Availability details for
Quantity
  • Availability
  • Add to Cart