ENA346974762 Enamine

decahydroquinoline-2-carboxylic acid hydrochloride

  • MDL number MFCD20731237
  • Cas Number : 253136-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-96545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96545
Availability details for
Quantity
  • Availability
  • Add to Cart