ENA346974764 Enamine

2-hydroxy-3-(4-methylphenyl)butanoic acid

  • MDL number MFCD20731239
  • Cas Number : 1354961-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96576

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96576
Availability details for
Quantity
  • Availability
  • Add to Cart