ENA346974766 Enamine

3-(aminomethyl)-3-ethylpentan-1-ol

  • MDL number MFCD20731242
  • Cas Number : 1354954-41-8
  • Molecular Weight : 145.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-96597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96597
Availability details for
Quantity
  • Availability
  • Add to Cart