ENA346974778 Enamine

tert-butyl 4-amino-4-methylpentanoate hydrochloride

  • MDL number MFCD20731256
  • Cas Number : 1354962-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-96705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96705
Availability details for
Quantity
  • Availability
  • Add to Cart