ENA346974782 Enamine

1-(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)-2,2,2-trifluoroethan-1-amine hydrochloride

  • MDL number MFCD20731263
  • Cas Number : 1354949-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96742
Availability details for
Quantity
  • Availability
  • Add to Cart