ENA346974793 Enamine

methyl 2-amino-4-bromo-3-methylbenzoate

  • MDL number MFCD17244231
  • Cas Number : 851045-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94720

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-94720
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart