ENA346974803 Enamine

2-(3-bromophenyl)-2-(pyrrolidin-1-yl)acetic acid hydrochloride

  • MDL number MFCD20731120
  • Cas Number : 1354949-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94819

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94819
Availability details for
Quantity
  • Availability
  • Add to Cart