ENA346974806 Enamine

3-methyl-1-(pyridin-4-yl)-1H-pyrazol-5-amine

  • MDL number MFCD06254124
  • Cas Number : 3524-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-94841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94841
Availability details for
Quantity
  • Availability
  • Add to Cart