ENA346974811 Enamine

ethyl 2-(ethylamino)-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD03674408
  • Cas Number : 4280-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94892
Availability details for
Quantity
  • Availability
  • Add to Cart