ENA346974814 Enamine

2-(5-bromothiophen-2-yl)propanoic acid

  • MDL number MFCD14530844
  • Cas Number : 139547-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94896

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94896
Availability details for
Quantity
  • Availability
  • Add to Cart