ENA346974817 Enamine

methyl 3-[5-(2,2-dimethylpropyl)-1,2,4-oxadiazol-3-yl]benzoate

  • MDL number MFCD20731125
  • Cas Number : 1354959-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94901
Availability details for
Quantity
  • Availability
  • Add to Cart