ENA346974819 Enamine

3-[(cyclopropylmethoxy)methyl]-4-fluorobenzoic acid

  • MDL number MFCD17287618
  • Cas Number : 1343367-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94903

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94903
Availability details for
Quantity
  • Availability
  • Add to Cart