ENA346974832 Enamine

4-[(3-methylbut-2-enamido)methyl]benzoic acid

  • MDL number MFCD11202664
  • Cas Number : 1038335-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94938

Safety Information

msds https://www.enaminestore.com/catalog/EN300-94938
Availability details for
Quantity
  • Availability
  • Add to Cart