ENA346974851 Enamine

2,2-dimethylbutan-1-ol

  • MDL number MFCD00021983
  • Cas Number : 1185-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95075

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95075
Availability details for
Quantity
  • Availability
  • Add to Cart