ENA346974871 Enamine

2-imino-5-methyl-1,3-thiazolidin-4-one

  • MDL number MFCD01332593
  • Cas Number : 3805-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95101

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95101
Availability details for
Quantity
  • Availability
  • Add to Cart