ENA346974874 Enamine

1-amino-2,3-diphenylpropan-2-ol

  • MDL number MFCD17387781
  • Cas Number : 126376-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95105

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95105
Availability details for
Quantity
  • Availability
  • Add to Cart