ENA346974876 Enamine

5-(3-cyanophenyl)-1,2,4-oxadiazole-3-carboxylic acid

  • MDL number MFCD18344108
  • Cas Number : 1342223-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95107

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95107
Availability details for
Quantity
  • Availability
  • Add to Cart