ENA346974882 Enamine

1-[2-(2-aminoethoxy)ethoxy]butane

  • MDL number MFCD11196423
  • Cas Number : 6338-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95147

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95147
Availability details for
Quantity
  • Availability
  • Add to Cart