ENA346974890 Enamine

3-[3-(2-oxopiperidin-1-yl)propoxy]benzonitrile

  • MDL number MFCD12454194
  • Cas Number : 1095101-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95158
Availability details for
Quantity
  • Availability
  • Add to Cart