ENA346974892 Enamine

N-butyl-4-chloro-2-hydroxybenzamide

  • MDL number MFCD00865617
  • Cas Number : 575-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-95161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95161
Availability details for
Quantity
  • Availability
  • Add to Cart