ENA346974894 Enamine

4-chloro-5-methylquinazoline

  • MDL number MFCD09998953
  • Cas Number : 90272-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-106
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67938
Availability details for
Quantity
  • Availability
  • Add to Cart