ENA346974900 Enamine

1-[3-(2-methoxyphenyl)-1,2,4-oxadiazol-5-yl]ethan-1-amine hydrochloride

  • MDL number MFCD20731145
  • Cas Number : 1354962-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95170

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95170
Availability details for
Quantity
  • Availability
  • Add to Cart