ENA346974901 Enamine

1-[3-(3-methoxyphenyl)-1,2,4-oxadiazol-5-yl]ethan-1-amine hydrochloride

  • MDL number MFCD20731146
  • Cas Number : 1354961-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149...151
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95171

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95171
Availability details for
Quantity
  • Availability
  • Add to Cart