ENA346974908 Enamine

3-(3,5-dichlorophenoxy)propanoic acid

  • MDL number MFCD09704798
  • Cas Number : 938118-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95235

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95235
Availability details for
Quantity
  • Availability
  • Add to Cart