ENA346974909 Enamine

2-[2-(4-methyl-1,3-thiazol-5-yl)-1H-1,3-benzodiazol-1-yl]acetic acid

  • MDL number MFCD12741857
  • Cas Number : 1094744-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95236

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95236
Availability details for
Quantity
  • Availability
  • Add to Cart