ENA346974911 Enamine

N,N-diethylpyrrolidin-3-amine

  • MDL number MFCD02184351
  • Cas Number : 20984-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68733

Safety Information

signalword Danger
pcodes P210,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P370+P378,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-68733
pictograms GHS05
hstatements H227,H314
Availability details for
Quantity
  • Availability
  • Add to Cart