ENA346974915 Enamine

1,2-dimethyl-1H-1,3-benzodiazol-5-amine

  • MDL number MFCD03872210
  • Cas Number : 3527-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95247

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95247
Availability details for
Quantity
  • Availability
  • Add to Cart