ENA346974917 Enamine

1,3-dimethyl-5-(2,3,5-trimethylphenoxy)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD13381821
  • Cas Number : 1152565-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95251
Availability details for
Quantity
  • Availability
  • Add to Cart