ENA346974922 Enamine

4-propylpyrrolidin-2-one

  • MDL number MFCD15146418
  • Cas Number : 89895-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 20-21
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-95272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95272
Availability details for
Quantity
  • Availability
  • Add to Cart