ENA346974929 Enamine

5-(trifluoromethyl)-1,3-benzothiazole-2-thiol

  • MDL number MFCD01809822
  • Cas Number : 23420-87-3
  • Molecular Weight : 235.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(trifluoromethyl)-1,3-benzothiazole-2(3H)-thione
Inchi Code 1S/C8H4F3NS2/c9-8(10,11)4-1-2-6-5(3-4)12-7(13)14-6/h1-3H,(H,12,13)
Inchi Key DJRUWESABPZMRB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-23811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23811
Availability details for
Quantity
  • Availability
  • Add to Cart