ENA346974932 Enamine

2-(cyclopentylformamido)-3-hydroxybutanoic acid

  • MDL number MFCD09836895
  • Cas Number : 1160699-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95303
Availability details for
Quantity
  • Availability
  • Add to Cart