ENA346974933 Enamine

4-(1,3-dioxaindan-5-yl)-2-methyl-5-propyl-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD20731158
  • Cas Number : 1354960-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95306
Availability details for
Quantity
  • Availability
  • Add to Cart