ENA346974937 Enamine

methyl 3-[(oxan-4-yl)amino]propanoate

  • MDL number MFCD12087741
  • Cas Number : 1137213-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95372

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95372
Availability details for
Quantity
  • Availability
  • Add to Cart