ENA346974940 Enamine

1-(4-fluorophenyl)-3-methylbutan-2-amine

  • MDL number MFCD11934946
  • Cas Number : 1178757-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95376

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95376
Availability details for
Quantity
  • Availability
  • Add to Cart