ENA346974941 Enamine

N-[4-(4-propylpiperazin-1-yl)pentyl]cyclopropanamine

  • MDL number MFCD17341399
  • Cas Number : 1284821-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95377
Availability details for
Quantity
  • Availability
  • Add to Cart