ENA346974944 Enamine

2-(4-bromothiophen-2-yl)-2-{[(oxolan-2-yl)methyl]amino}acetic acid hydrochloride

  • MDL number MFCD20731161
  • Cas Number : 1354949-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95384
Availability details for
Quantity
  • Availability
  • Add to Cart