ENA346974962 Enamine

2-[(phenylamino)methyl]aniline

  • MDL number MFCD14597920
  • Cas Number : 20887-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95457

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-95457
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart