ENA346974964 Enamine

4-ethenyl-2,6-dimethoxyphenol

  • MDL number MFCD02094170
  • Cas Number : 28343-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95459

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95459
Availability details for
Quantity
  • Availability
  • Add to Cart