ENA346974986 Enamine

4-(2H-1,3-benzodioxol-5-yl)-5-cyclopentyl-2-methyl-2,3-dihydro-1H-pyrazol-3-imine

  • MDL number MFCD13660328
  • Cas Number : 1356725-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-95504

Safety Information

msds https://www.enaminestore.com/catalog/EN300-95504
Availability details for
Quantity
  • Availability
  • Add to Cart